To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Tag
- Antigen name:GST Tag
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:S. japonicum
- Conjugation:Unconjugated
- ELISA:Yes
- Size:40 μg
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:Supplied in PBS, pH 7.4 with 0.02% sodium azide
- Storage temperature:The product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
- Concentration:Reconstitute the lyophilized antibody with deionized water (or equivalent) to the final concentration of 0.5 mg/ml
- Immunogen:Recombinant Glutathione-S-Transferase from Schistosoma japonicum
- Cat. no.:77007-622
- Packaging:Vial
Spécifications
A propos de cet article
GenScript Rabbit Anti-GST-tag Polyclonal Antibody is highly purified from rabbit antiserum by immunoaffinity chromatography. The immunogen used is a recombinant version of glutathione-S-transferase from Schistosoma japonicum. Glutathione-S-transferase is an epitope tag in generating fusion proteins. Antibodies specific to fusion proteins are useful in monitoring protein expression in Western Blots as well as immunoaffinity purification of proteins expressed in E. coli. The glutathione-S-transferases are a family of enzymes that catalyze the addition of the tripeptide glutathione to endogenous and xenobiotic substrates which have electrophilic functional groups. They play an important role in the detoxication and metabolism of many xenobiotic and endobiotic compounds. The glutathione adducts produced have increased solubility in water and are subsequently enzymatically degraded to mercapturates and excreted.
- High specificity
- High affinity
- High stability
GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.
Type: Tag
Antigen: GST Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: S. japonicum