Nemiana simplex Replica
# de catalogue 470128-300
Fournisseur: PARKER PALEONTOLOGICAL ENT.
Spécifications
- Description:Nemiana simplex replica
- Cat. no.:470128-300
- Supplier No.:PPC-47
Spécifications
A propos de cet article
Beautifully remastered fossil replica. Great for paleontological identification, lectures on evolution, and display.
- Exact replica
- Plastic resin
- 4×3" plaque
Nemiana simplex may represent siphonocladalean chlorophyte algae on the underside of micaceous quartzose sandstone slabs from the Precambrian of Ukraine. Such green algae are commonly called 'ball or bubble algae'. So, Nemiana may not be an animal at all. Nemiana is found from the Yampol Member, Mogilev Formation, Balday Series, upper Vendian (= upper Ediacaran), upper Neoproterozoic of Novodnestrovsky Quarry, Podolia, Ukraine. The ediacaran biota existed from about 580 to 540 million years ago.
Customers Who Bought This Also Bought