ContactUsLinkComponent
PTS Collect™ Capillary Tubes 15 μl
PTS Collect™ Capillary Tubes 15 μl
Catalog # 77020-356
Supplier:  PTS Diagnostics
New Product
PTS COLLECNT CAPILLARY TUBES 15UL 25CNT
PTS Collect™ Capillary Tubes 15 μl
Catalog # 77020-356
Supplier:  PTS Diagnostics
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    PTS Collect™ capillary tubes, 15 μl
  • Cat. no.:
    77020-356
Specifications
About this item
PTS Collect™ capillary tubes provide speed, efficiency and savings — especially for applications where exact sample volumes are required.
  • Ease-of-use with no assembly
  • One-hand blood collection and dosing
  • Glass tube for easy blood collection
  • Bulb-squeeze dispensing technology that ensures consistent performance
  • Only one item to inventory (no plunger)
PTS Collect capillary tubes provide better efficiency and are designed to be easier to use than traditional blood transfer tubes. Using a single glass tube with plastic casing, PTS Collect capillary tubes provide consistent self-wicking technology to preset volumes. This improved design is perfect for blood collection needs covering a wide range of applications across many different medical devices.
Customers Who Bought This Also Bought