A propos de cet article
The Epredia Laser Cassette Printing technology produces high quality print combined with a patented chemical additive in the cassette to deliver consistency and efficiency. Built in interface connectivity ensures LIS/LIM compatibility.
- Epredia NOVA laser Printer is a single cassette hopper printer providing an economical option for laboratories
- Three additional hoppers are included for convenient cassette exchanges
- Epredia VEGA laser printer has six cassette hoppers and supports laboratories printing a variety of cassette colors and styles
- Laser Technology eliminates ancillary components such as print heads & ribbons and simplifies operation
- High resolution printing, 600 dpi, improves printing sharpness and clarity of characters allowing more characters to be embedded on cassette surface
- Fully automated system with fast print speed, five seconds per cassette, allows the user to quickly and easily print cassettes needed for a case at accessioning or grossing
- Laser system with cassette additive print technology ensures permanent and delible marking that are extremely durable overtime
- Small footprint reduces the concern of space constraint in the lab
- Quiet printing reduces distraction during grossing and accessioning
Workflow Efficiencies –
Epredia laser printers are designed to create crisp, accurate and indelible print quality for laboratories concerned about workflow bottlenecks caused by the need for reprinting cassettes with low-quality or unreadable printing at accessioning.
High Printing Quality –
•Epredia laser printers create sustainable and indelible printed information where patient data can be identified accurately throughout the lifetime of the cassette supporting laboratories and patients.
•Character Types: Alphanumeric.
•Barcode Types: Linear, Bidimensional, ASCII Codes.
High Throughput –
Epredia laser printers incorporate a highly efficient technology helping labs with higher printing demands utilize the laser printing capabilities and automated cassette feeder feature to achieve high throughput and faster turn-around times.
Low Operations Cost –
Epredia laser printers do not require replacement of ancillary components significantly decreasing operation costs and lowers any intensive maintenance helping lab managers from being burdened with high operation costs.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 76540-420
- 76540-418
- 76540-446
- 76540-444
- Compatibility:
- LIS/LIM
- LIS/LIM
- LIS/LIM
- LIS/LIM
- Print resolution:
- 600 dpi
- 600 dpi
- 600 dpi
- 600 dpi
- Width:
- 7.9"
- 10.2"
- 10.2"
- 7.9"
- Depth:
- 20.1"
- 24.4"
- 24.4"
- 20.1"
- Height:
- 20.5"
- 20.5"
- 20.5"
- 20.5"
- Weight:
- 36 lbs.
- 60 lbs.
- 60 lbs.
- 36 lbs.