VWR® Cryogenic Dot and Strip Labels
Supplier: VWR International
Certificates
About this item
VWR® Cryogenic dot and strip labels are ideal for all cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications.
- Can be placed in most incubators
- DYMO or Zebra compatible
- Direct thermal and cryogenic
Labels feature a unique adhesive that can withstand extreme temperatures from –196 to 121 °C (–321 to 250 °F), without the need for lamination.
Ordering information: Rolls of labels are supplied on a 44 mm Ø (1.75") core. Laser printer label sheets are 21.6W×27.9L cm (8.5×11"). Assorted labels come in blue, green, purple, red, and yellow. Roll of assorted labels contains 1000 labels of each color. Boxes of assorted laser printer labels contain five sheets of each color.
Customers Who Bought This Also Bought