VWR® Silicone Sealing Mats for VWR® Deep Well Plates
Fournisseur: SCIENTIFIC SPECIALTIES, INC.
New Product
Certificats
A propos de cet article
Provide a safe, secure seal to minimize sample loss with VWR® Deep Well Storage Plates.
- Reusable and pierceable
- For use in pharmaceutical industry, food industry, and molecular biology
- Temperature range: –80 to +120 °C
These mats are compatible with VWR® Deep Well Plates in the series: 76924-044 through 76924-058.
Certifications: Certified free of DNase, RNase, DNA and pyrogens. Sterile mats are also endotoxin-free.
Customers Who Bought This Also Bought