Replacement Air Filter for ULT Freezers
Catalog # 76779-128
Supplier: Thermo Fisher Scientific
Specifications
- Description:Replacement air filter
- Cat. no.:76779-128
- For Use With:ULT chest freezers
Specifications
About this item
Replacement air filter for ultra-low temperature chest freezers.
Routine maintenance is critical for proper ultra-low temperature (ULT) freezer operation. Replacing your ULT's old air filter enables the necessary airflow to the unit's refrigeration system.
Customers Who Bought This Also Bought