Bioreactor Complete Accessories Kit
# de catalogue 75780-186
Fournisseur: Chemglass
Spécifications
- Description:Bio reactor accessory kit complete, non sterile
- Cat. no.:75780-186
- Supplier No.:CLS-1390-01
Spécifications
A propos de cet article
This pre fabricated, single use bio reactor kit consists of the 8 components. They are packed in an ISO Class 7 clean room, double bagged and ready for use.
- Minimize labor costs
- Eliminates cleaning validation
- Reduces set up time and costs
- Minimize injuries associated with cutting in-house
- Improved consistency
- Gives the ability to assemble in open lab type environment
- Provides aseptic solution to close off an unused line
- Autoclave maxium 15 psi @ 134 °C, 60 min
- Gamma radiation 25 kGy to 50 kGy
Kits are fabricated with full traceability and accompanying validation documentation, which meets USP Class VI requirements. These pre-assembled kits can be customized to any validated components currently specified on your bioreactor design.
Customers Who Bought This Also Bought