VWR® Cleanroom Safety Shoes, Slip-On
Fournisseur: VWR International
New Product
Certificats
A propos de cet article
Premium comfort cleanroom and safety shoes.
- Comfortable, form fitting, and lightweight design
- Microfiber upper, with polyester fiber sewing
- Mesh interior for breathability
- Anti-static sole material
- Polycarbonate toe cap
Premium cleanroom shoes designed with foot comfort in mind. These shoes feature a microfiber PU leather upper that is 1.8 mm thick. The mesh interior provides added breathability. Polyester fiber seams.
Customers Who Bought This Also Bought