HyperSep™ SCX Strong Cation Exchanger SPE Plates
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
A strong cation sorbent for extraction of charged basic compounds.
- Particle size: 40 to 63 µm
- Pore size: 60 Å
- Surface area: 525 m²/g
Customers Who Bought This Also Bought