HyperSep™ Silica SPE Plates
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
A polar sorbent primarily used to retain analytes in non-polar matrices.
- Particle size: 40 to 63 µm
- Pore size: 60 Å
- Surface area: 530 m²/g
Customers Who Bought This Also Bought