HyperSep™ Phenyl SPE Plates
# de catalogue 10047-810
Fournisseur: Thermo Scientific
Spécifications
- Volume:1 ml
- Description:Phenyl phase plates, bed weight 100 mg
- Cat. no.:10047-810
- Supplier No.:60300-688
Spécifications
A propos de cet article
HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation.
- Particle size: 40 to 60 µm
- Pore size: 61 Å
- Surface area: 481 m²/g
Well plates are customisable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing.
Customers Who Bought This Also Bought