HyperSep™ Aminopropyl SPE Plates
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
A polar sorbent for both polar and anion exchange interactions.
- Particle size: 40 to 63 µm
- Pore size: 58 Å
- Surface area: 496 m²/g
Customers Who Bought This Also Bought