HyperSep™ Hypercarb SPE Plates
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Unique material for retention of highly polar compounds.
- Particle size: 30 to 40 µm
- Pore size: 150 to 300 Å
- Surface area: 108 to 148 m²/g
Customers Who Bought This Also Bought