miniPCR® miniX Thermal Cyclers
Fournisseur: AMPLYUS LLC
New Product
Certificats
A propos de cet article
These 8 or 16-well thermal cyclers offer a personalized PCR experience for maximum engagement at a budget-conscious price point. Small and portable, this fully-featured PCR machine fits on your bench and travels wherever your science goes.
- Peer-reviewed published results
- Durable in the lab, classroom, field, even outer space
- Weighs under one pound, fits in the palm of your hand
- AC or battery powered (battery not included)
- Universal compatibility via USB or Bluetooth®
- 3-year warranty and 30-day satisfaction guarantee
- Fully compatible with standard PCR tubes and reagents
Integrated with the latest technology and designed for peak performance with drop-proof durability, these are the ultimate thermal cyclers for classroom or field work.
Customers Who Bought This Also Bought