ContactUsLinkComponent
miniPCR® Plant Genetics Lab: Taking Mendel Molecular with Wisconsin Fast Plants®
miniPCR® Plant Genetics Lab: Taking Mendel Molecular with Wisconsin Fast Plants®
# de catalogue 470351-998
Fournisseur:  AMPLYUS LLC
New Product
MINIPCR PLANT GENETICS LAB
miniPCR® Plant Genetics Lab: Taking Mendel Molecular with Wisconsin Fast Plants®
# de catalogue 470351-998
Fournisseur:  AMPLYUS LLC
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    miniPCR® plant genetics lab
  • No. of Lab Groups:
    8
  • Cat. no.:
    470351-998
Spécifications
A propos de cet article
Investigate the genotypic basis of a phenotype using Wisconsin Fast Plants®.
  • Techniques: Micropipetting, DNA extraction, PCR, gel electrophoresis
  • Topics: Genetic inheritance, genotype to phenotype, plant biotechnology
  • Time required: Can be completed in two 45-minute class periods or one 120-minute class period
  • Level: General high school through college
Investigate the genotypic basis of an observable phenotype using Rapid Cycling Brassica rapa (RCBr), also known by the trademark name Wisconsin Fast Plants®. Wild-type plants grow with a distinctive purple stem due to the pigment anthocyanin. In some RCBr plants, a mutation disrupts the anthocyanin production pathway and leads to green stems. The gene responsible for the purple vs. green color is named Anthocyaninless because the mutant form leads to decreased production of the pigment anthocyanin. Test the Anthocyaninless gene from different plants to link genotype to phenotype.
Reagents and consumables supplied by user:
- Agarose (electrophoresis grade)
- DNA stain (e.g., GelGreen™)
- Gel electrophoresis buffer (e.g., 1X TBE)
- Distilled or deionized H₂O (to dilute buffer concentrate – not included in Lab Companion Kit)
- Microtubes
- PCR tubes
Required equipment:
- Micropipettes and tips
- Thermal cycler
- Gel electrophoresis and visualization system
BodyTextAccessories: The learning lab companion kit (catalog no. 470352-026) contains all the consumables (except water) needed for a class of 32 students. These reagents and consumables can also be sourced independently.
Delivery: The miniPCR Plant Genetics Lab kit contains reagents for 8 lab groups: 5X EZ PCR Master Mix, Load Ready™; Plant Genetics Lab Primer Mix; DPX Buffer 100 bp DNA Ladder, Load Ready™. Note: Reagents are compatible with blueGel™ or other standard horizontal electrophoresis systems.
Customers Who Bought This Also Bought