ContactUsLinkComponent
miniPCR® Dye Electrophoresis Lab: Mendel’s Peas
miniPCR® Dye Electrophoresis Lab: Mendel’s Peas
# de catalogue 470352-034
Fournisseur:  AMPLYUS LLC
New Product
MINIPCR MENDELS PEAS LAB
miniPCR® Dye Electrophoresis Lab: Mendel’s Peas
# de catalogue 470352-034
Fournisseur:  AMPLYUS LLC
New Product
Spécifications
  • Description:
    miniPCR® dye electrophoresis lab: Mendel's Peas
  • No. of Lab Groups:
    8
  • Cat. no.:
    470352-034
Spécifications
A propos de cet article
Some of Mendel’s peas have been discovered in an old notebook!
  • An engaging and accessible introduction to gel electrophoresis
  • Safe, non-toxic dyes that are directly visible without gel staining
  • Pre-aliquoted samples and premeasured agarose tablets for easy prep
  • Gel electrophoresis reagents included: TBE electrophoresis buffer and 8 Agarose Tabs™
  • Compatible with Bandit™, blueGel™, and traditional gel electrophoresis systems
  • A low-cost jumping-off point to explore microbiology, biotechnology, and molecular identification!
This dye electrophoresis lab connects traditional Mendelian genetics with our modern understanding of DNA and inheritance. Students play the role of plant geneticists testing peas from Mendel’s notebook. Gel electrophoresis is used to analyze simulated DNA samples from Mendel’s peas to examine how genetic differences control pea shape.
Supplied by user:
- Distilled or deionized H₂O (to dissolve TBE buffer)
- Gel electrophoresis system (lab is compatible with Bandit™, blueGel™, or other gel electrophoresis systems)
- Variable volume micropipettes, or fixed-volume minipettes, and tips
Required equipment:
- 2 to 20 μl variable volume or 10 μl fixed volume micropipettes and tips
- Gel electrophoresis system (e.g., Bandit™, blueGel™, or other gel electrophoresis systems)
Storage notes:
- Reagents must be used within 12 months of shipment
- Agarose and buffer can be stored at room temperature
- Refrigeration is recommended for blue dye samples to prevent evaporation
Note: This lab is compatible with any horizontal gel electrophoresis system.
BodyTextAccessories: The learning lab companion kit (catalog no. 470352-026) contains all the consumables (except water) needed for a class of 32 students. These reagents and consumables can also be sourced independently.
Delivery: This lab kit provides enough reagents for eight lab groups: 8 Load Ready™ Strips (tube strips containing pre-aliquoted dye samples); TBE electrophoresis buffer; 8 Agarose Tabs™.
Customers Who Bought This Also Bought