VWR® UHP Multichannel Pipettes
Supplier: VWR International
Certificates
About this item
The VWR® UHP multichannel pipette is available in sizes from 10 to 300 µl with a lightweight and durable design. These pipettes will provide you with low pipetting forces and the highest levels of accuracy and precision.
- Autolocking volume mechanism to avoid volume drift while pipetting
- Fully autoclaveable and UV resistant
- Color coded ejector button and calibration switch make for easy identification
- Universal fit with most common brands of pipette tips
- Lightweight design, quality materials, low pipetting forces, and a 5-ear warranty
The VWR® multichannel pipette is calibrated to ISO 8655 standards using VWR® Universal pipette tips. Pipettes were tested for both autoclave and UV resistance.
Certifications: In compliance with ISO 8655.
BodyTextAccessories: Pipette Stands are available in the accessories tab below.
Packaging: Includes a QC certificate, pipette, and a quick reference guide.
Customers Who Bought This Also Bought