miniPCR® Silicone Practice Gels, Set of 10
# de catalogue 470352-018
Fournisseur: AMPLYUS LLC
New Product
Spécifications
- Description:miniPCR® silicone practice gels, set of 10
- Cat. no.:470352-018
Spécifications
A propos de cet article
Set of 10 durable, reusable silicone practice gels.
Hone your gel loading skills with this set of 10 durable, reusable silicone practice gels. These are the same reusable gels included with the Micropipetting 101 Learning Lab.
BodyTextAccessories: The Learning Lab Companion Kit (catalog no. 470352-026) contains all the consumables (except water) needed for a class of 32 students. These reagents and consumables can also be sourced independently.
Delivery: Includes: 10 silicone practice gels (6×6 cm each); top row loading: 9 wells, for everyday practice; bottom row loading: 13 wells, for advanced practice.
Customers Who Bought This Also Bought