VWR® Cleanroom Gloves, Latex, Non Sterile, Class 100
Fournisseur: VWR International
Certificats
A propos de cet article
Ambidextrous, powder-free gloves designed to provide comfort and tactile sensitivity.
- Ambidextrous, beaded cuff
- Powder free, silicone free
- Natural rubber latex
- Low particle count
- Specialized manufacturing process
VWR® cleanroom gloves, latex, non sterile, 12", Class 100.
Spécifications
- Catalog No:
- 76579-686
- 76579-680
- 76579-684
- 76579-682
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile