miniPCR® Bandit™ Biotech Bundle
# de catalogue 470352-074
Fournisseur: AMPLYUS LLC
New Product
Spécifications
- Description:Bandit™ Biotech bundle
Spécifications
A propos de cet article
Get hands-on with biotech fundamentals! With the Bandit™ Biotech Bundle you will have everything you need to:
- Build a functional and reusable gel electrophoresis system
- Discover how circuits and electric fields can be used to investigate the properties of molecules
- Develop pipetting skills by engaging your creative side with pipette art
- Explore genetics concepts with associated Dye Electrophoresis Labs (sold separately)
Teach fundamental biotech skills and methods while illuminating underlying physical and chemical principles. Then, apply those techniques to answer biological questions. All on a classroom-friendly budget!
Ordering information: USB-C power source required for operation is not included.
Delivery: Bandit™ Biotech Bundle includes: Bandit™ STEM Electrophoresis Kit: Build it to understand it! Perfect for introducing the fundamentals of gel electrophoresis to introductory users. 10 µl Minipette: Fixed volume micropipette perfect for learning to pipette and load gels. Bag of 100 micropipette tips 2 to 200 µl: Enough tips to get you going. Micropipetting Practice Dyes: Hone your pipetting skills with 5 ml each of blue, yellow and red micropipetting practice dyes. Micropipette Art Card: Use your pipetting dyes to put the A in STEAM. This reusable card pairs with our Micropipette Art activity so you can create colorful, artistic images all while developing your pipetting skills.
Customers Who Bought This Also Bought