miniPCR® Premium Classroom Pack
# de catalogue 470352-078
Fournisseur: AMPLYUS LLC
New Product
Spécifications
- Description:miniPCR® Premium classroom pack
Spécifications
A propos de cet article
Bring a bonanza of cutting-edge biotechnology into your classroom with the miniPCR® Premium Classroom Pack.
Maximize hands-on learning with this Premium Classroom Pack from miniPCR bio™. Ideal for classes between 32 and 64 students working in groups of 2 to 4 students.
Certifications: The miniPCR thermal cyclers and blueGel electrophoresis systems are CE marked, made in the USA, and backed by the miniPCR 2-year Warranty and 30-day satisfaction guarantee!
Delivery: This package includes: 8 miniPCR® mini8X thermal cyclers: Simple and intuitive, the mini8 offers a personalized PCR experience for maximum engagement. 8 blueGel™ electrophoresis systems: Fast, durable, safe, and easy to use, blueGel™ allows you to visualize results instantly in a convenient, all-in-one device compatible with safe DNA dyes such as GelGreen®. 2 Gyro™ Plus Microcentrifuges: This robust, variable speed microcentrifuge is user friendly and includes rotors for both 1.5 ml tubes and 0.2 ml tubes. Micropipetting supplies: Our H-style micropipettes are durable, accurate, and fully adjustable. Pack includes: 16 student micropipettes (2 to 20 μl), 1 teacher set of 4 micropipettes (0.5 to 10 μl, 2 to 20 μl, 20 to 200 μl, 100 to 1000 μl), compatible tips (8 racks 200 μl tips, 2 racks 10 μl tips, 2 racks 1000 μl tips), and 2 micropipette stands for storage.
Customers Who Bought This Also Bought