miniPCR® Starter Classroom Pack
# de catalogue 470352-068
Fournisseur: AMPLYUS LLC
New Product
Spécifications
- Description:miniPCR® Starter classroom pack
- Cat. no.:470352-068
Spécifications
A propos de cet article
Bring cutting-edge biotechnology into your classroom with the miniPCR® Starter classroom pack.
Ideal for classes between 16 and 32 students working in 4 to 8 lab groups.
Certifications: The miniPCR® thermal cyclers and blueGel electrophoresis systems are CE marked, made in the USA, and backed by the miniPCR 2-year Warranty and 30-day satisfaction guarantee!
Delivery: This Starter Classroom Pack includes: 2 miniPCR® mini16X thermal cyclers: Featuring 16-well capacity, USB and Bluetooth connectivity, mini16 is broadly compatible with computers and smartphones including iPads and iPhones. 4 blueGel™ electrophoresis systems: Fast, durable, safe, and easy to use, blueGel™ allows you to visualize results instantly in a convenient, all-in-one device compatible with safe DNA dyes such as GelGreen®. 1 Gyro™ Microcentrifuge: This robust, single speed microcentrifuge is user-friendly and includes rotors for both 1.5 ml tubes and 0.2 ml tubes. Micropipetting supplies: Our H-style micropipettes are durable, accurate, and fully adjustable. Pack includes 4 student micropipettes (2 to 20 μl), 1 teacher set of 3 micropipettes (0.5 to 10 μl, 2 to 20 μl, 20 to 200 μl), and compatible tips (4 racks 200 μl tips, 2 racks 10 μl tips).
Customers Who Bought This Also Bought