Spécifications
- Description:HABITAT sample flask rack
- Cat. no.:76836-702
Spécifications
A propos de cet article
With the help of this stand, you can safely store your sample bottles.
There are four positions for 250 ml bottles. Due to a slightly inclined position, the sample bottle can be completely emptied without any problems.