ContactUsLinkComponent
HABITAT research Bioreactor, HABITAT ferment cell Control Unit Package
HABITAT research Bioreactor, HABITAT ferment cell Control Unit Package
# de catalogue 76836-712
Fournisseur:  IKA Works
FERMENT HABITAT
HABITAT research Bioreactor, HABITAT ferment cell Control Unit Package
# de catalogue 76836-712
Fournisseur:  IKA Works
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    HABITAT ferment
  • Cat. no.:
    76836-712
Spécifications
A propos de cet article
HABITAT research is the smart laboratory bioreactor from IKA. As the first bioreactor with a lid stand, it ensures ergonomic working and a well-organized laboratory.
  • 4 peristaltic pumps allow easy tube loading, variable speed, bi-directional and speed feedback control, volume and flowrate control
  • Includes user-friendly software with functional modes like batch, fed-batch, perfusion, chaotic mixing, adjustable PID
  • New chaotic mixing option ensures a more homogeneous mixture
  • Minimal footprint to save working space
To operate HABITAT research, you only need a control unit package and a vessel package. For sensitive cell cultures such as mammalian cells, select the cell variant, with gassing rates from 0 to 2000 cc/min. If you are using HABITAT as a fermenter, we recommend the ferment variant, with gassing rates of 0 to 20000 cc/min. If you decide to use the photobioreactor, the photo version with LED panels is perfect for you. The suffix 'cct' in a package indicates that there are connection options for three additional sensors: turbidity, CO₂ and conductivity. The second step is to complement your control unit package with the vessel package that matches your application and working volume.
Certifications: CE.
Ordering information: 1+1 Year warranty.
Delivery: Supplied with: HABITAT photo ferment S099, tablet with software for bioreactor and HA.pl. Peltier cooling element.
Customers Who Bought This Also Bought