ContactUsLinkComponent
Liquid Chromatograph with UV Detector
Liquid Chromatograph with UV Detector
# de catalogue 76836-906
Fournisseur:  LUCIDITY SYSTEMS LLC
New Product
LUCIDITY CHROMATOGRAPH LIQUID LC-UV
Liquid Chromatograph with UV Detector
# de catalogue 76836-906
Fournisseur:  LUCIDITY SYSTEMS LLC
New Product
Spécifications
  • Description:
    High performace liquid chromatograph with UV detector, 14×20×17"
Spécifications
A propos de cet article
High Performance Liquid Chromatograph with UV detector.
  • LC-UV with integrated multi-wavelength UV detector
  • Equipped with a 20 place autosampler
  • Binary pump system
  • High pressure HPLC pumps for gradient flow up to 10000 psi
  • Integrated degassers and temperature programmable column oven
  • Includes laptop with Lucidity Software
The Lucidity LC-UV is one of the smallest, most economical, and robust HPLCs on the market. Equipped with a variable wavelength UV detector capable of measuring signal from 185 nm to 475 nm, the Lucidity LC-UV has a binary gradient pumping system existing of two high pressure piston pumps that deliver accurate solvent gradients up to 10000 psi.
With a 20-position autosampler and integrated degassers the Lucidity LC-UV has everything required to provide great chromatography results.
Delivery: The instrument comes with everything required for analysis, including: laptop containg Lucidity Software, starter column, full accessory kit and training videos. Self-installable in under 30 minutes. With a 24-month warranty, the LC-UV will provide reliable results for years to come.
Customers Who Bought This Also Bought