GC-FID Autosampler
# de catalogue 76836-904
Fournisseur: LUCIDITY SYSTEMS LLC
New Product
Spécifications
- Description:Lucidity Gas chromatograph autosampler
- Cat. no.:76836-904
Spécifications
A propos de cet article
GC-FID 16 place autosampler.
- Self-aligning autosampler
- Quick and easy syringe changing capabilities
- 16-position turntable
The Lucidity GC-FID autosampler installs quickly on the GC-FID with no alignment required. Autosampler has a 16-place turn-table that allows for multiple samples to be processed automatically. Easy access allows for quick vial placement and contant visibility of sampling. The autosampler is one of the smallest and lowest cost options for automated sampling.
Customers Who Bought This Also Bought