Gas Chromatograph with FID (GC-FID)
Fournisseur: LUCIDITY SYSTEMS LLC
New Product
Certificats
A propos de cet article
Compact, economical, fully functional gas chromatograph.
- Lucidity quick column change for quick and easy column installation
- Equipped with a split/splitless injection port
- Contains a very small oven which allows for rapid heating and cooling
- Intuitive Lucidity software included with purchase of instrument
- Instrument changes color to indicate status
- Self-installable and ease of use design
- Low power consumption
The Lucidity GC has one of the smallest footprints of any fully functional GC-FID. Changing GC columns in the Lucidity GC is as simple as loading a gaming cartridge. Fused silica and metal capillary columns can be installed into the column holder.
Delivery: The instrument comes with everything required for analysis, including: laptop containg Lucidity Software, starter column, full accessory kit and training videos. Self-installable in under 30 minutes. With a 24-month warranty, this GC-FID will provide reliable results for years to come.
Customers Who Bought This Also Bought