Virus Recombinant SARS-CoV-2 Omicron BA.4/5 Spike Protein Receptor Binding Domain (RBD) (from Insect cell), His Tag
Fournisseur: ReVacc Scientific
Certificats
A propos de cet article
A recombinant form of the receptor binding domain (RBD) with amino acids 319 to 541) of the spike (S) glycoprotein gene from SARS-CoV-2 bearing mutations identified in variant of Omicron BA.4/5, was produced by insect cells, followed by purification.
- This product contains C-terminal 6 hexa-histidine tag
This product has been run on SDS-PAGE with reduced condition. We also evaluated its binding to ACE2-Fc, anti-His mAb and a negative mAb in ELISAs.
Customers Who Bought This Also Bought