Virus Recombinant SARS-CoV-2 Nucleocapsid (N) Protein – Full length (from Insect cell), His Tag
Fournisseur: ReVacc Scientific
Certificats
A propos de cet article
A recombinant form of nucleocapsid (N) protein full length (AA 1-419) from SARS-CoV-2 virus (Wuhan Strain, GenBank: NC_045512.2) was produced by insect cells, followed by purification. This product contains C-terminal 6 hexa-histidine tag.
This product has been run on SDS-PAGE with reduced condition. We evaluated the binding of this protein to mAb clone RV20 (Cat# W89S59), two tag-specific antibodies (anti-His or Myc) and one convalescent serum in ELISAs.
Customers Who Bought This Also Bought