To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Tocilizumab
- Clonality:Monoclonal
- Clone:6C10
- Host:Mouse
- Isotype:IgG2b
- Antigen symbol:Tocilizumab
- Conjugation:Unconjugated
- ELISA:Yes
- Size:40 µg
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS, pH 7.4, containing 0.02% sodium azide
- Storage temperature:The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid freeze/thaw cycles
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
- Shipping temperature:Ice
- Immunogen:Tocilizumab
- Purification:Protein A affinity column
- Cat. no.:76509-984
- Packaging:Vial
Spécifications
A propos de cet article
The product is specific for tocilizumab. The antibody is recommended as a detection antibody in a pharmacokinetic (PK) bridging assay with capture antibody GenScript, A01978-40, anti-tocilizumab antibody (14B10), mAb, mouse.
- Anti idiotype antibodies
- High specificit
- High affinity
- High stability
GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.
.
Type: Primary
Antigen: Tocilizumab
Clonality: Monoclonal
Clone: 6C10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: