Stand and Holder for HandyStep® touch Pipette, BrandTech
Fournisseur: Brandtech
Certificats
A propos de cet article
This stand for the HandyStep® touch pipette securely holds the device, even with a PD-Tip II inserted, and saves space on the laboratory bench. It can be used for both the HandyStep® touch and the HandyStep® touch S.
Holder can be attached to a shelf or to a wall (adhesive back).
Customers Who Bought This Also Bought