Reagent Reservoirs, Gilson
Supplier: GILSON, INC.
Certificates
About this item
These reagent reservoirs are ideal for dispensing reagents and solutions using a multichannel pipette. The disposable, white PS reservoirs are available in 25-mL and 50-mL sizes.
- Wide stable bases, angled sides, and V-shaped bottoms allow for efficient and nearly complete removal of solutions
- Reservoirs provide spouts on all corners to simplify emptying and reduce spillage when removing liquids
- Middle volume graduations on the inside walls for quick measurement of liquids within the trough
Customers Who Bought This Also Bought