Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Rat Granzyme B ELISA Kit | Avantor

Rat Granzyme B ELISA Kit

Rat granzyme B ELISA kit, 96 tests
Catalog # 76730-944
Supplier:   Antibodies.com
Fournisseur #:  A80022-96
Rat Granzyme B ELISA Kit
Catalog # 76730-944
Supplier:  Antibodies.com
CAS Number:  
Rat Granzyme B ELISA Kit
Rat Granzyme B ELISA Kit

Rat granzyme B ELISA kit, 96 tests

CAS Number:  
Price
$1,071.04
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Rat Granzyme B ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Granzyme B in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 1.563 - 100 pg/ml
  • Sensitivity: 0.938 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Rat Granzyme B ELISA kit (A80022) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Granzyme B in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Granzyme B has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Granzyme B present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Granzyme B Antibody, which binds the captured Granzyme B present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Granzyme B captured in each well. The concentration of Granzyme B can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Spécifications

Spécifications

  • Désignation:
    Rat granzyme B ELISA kit
  • Taille:
    96 tests
  • Primary antibody reactivity:
    Rat
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Sample volume:
    100 μl
  • Detection range:
    1.563 - 100 pg/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    4 h 30 min
  • Cross reactivity:
    Rat Granzyme B ELISA kit exhibits high specificity and excellent specificity for the detection of rat Granzyme B. No significant cross-reactivity or interference between Granzyme B and analogues was observed.
  • Assay Principle:
    Quantitative
  • Cat. No.:
    76730-944
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    0.938 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    Granzyme B
  • Regulatory status:
    RUO

Spécifications