A propos de cet article
ABS Premium Laboratory Refrigerators deliver next level cooling to laboratory environments.
- Energy saving VSC compressors
- Microprocessor temperature controllers
- Ultra-quiet operation
- Fully array of alarms
- Controller battery backup
- Natural hydrocarbon refrigerants
- ENERGY STAR certified*
Engineered with variable speed compressors (VSCs), these units feature ultra-quiet operation and significant energy savings. VSCs optimize energy consumption by self-adjusting to cooling demands. These compressors also deliver enhanced system performance and provide a longer lifespan than other compressor variations.
Enjoy the uniformity and speed of microprocessor temperature control and a full array of alarms to safeguard your products. Unit controllers also come with a battery backup to keep data safe. Upgrade your laboratory environment with these premium refrigerators and welcome energy savings, noise reduction, smooth operation, and improved system performance in the work environment.
*Select models ENERGY STAR certified: 76808-814, 76808-816, 76808-822, and 76808-824
Spécifications
- Catalog No:
- 76808-816
- 76808-814
- 76808-824
- 76808-822
- Environmentally Preferable:
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Cat. No.:
- 76808-816
- 76808-814
- 76808-824
- 76808-822
- Defrost:
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Shelves/drawers:
- 10 adjustable shelves
- 5 adjustable shelves
- 10 adjustable shelves
- 5 adjustable shelves
- Door type:
- 2 glass swing doors
- 1 glass swing door, right hinged
- 2 solid swing doors
- 1 solid swing door, right hinged
- Weight:
- 264 kg (581 lbs.)
- 159 kg (350 lbs.)
- 223 kg (491 lbs.)
- 124 kg (274 lbs.)