Quarter-Watt Resistors
Fournisseur: MOUSER ELECTRONICS SE
Certificats
A propos de cet article
Quality resistors for electrical experiments.
- Quality carbon resistors
- Power rating: 0.25 Watts
- Color coded resistance values
- 20 to 25 mm wire leads
These resistors are designed to easily fit into protoboards or breadboards. Each 0.25 W resistor is color coded to designate its electrical resistance. The leads on each end are about 1" long.
Ordering information: These items are sold as individual resistors. Quantites greater than 1 may come taped at the ends.
Customers Who Bought This Also Bought