Eisco® 3 in 1 Weather Station
# de catalogue 470345-780
Fournisseur: Wards
Spécifications
- Description:3 in 1 weather station, 5.12" diameter
- Cat. no.:470345-780
Spécifications
A propos de cet article
5.12" diameter device for measuring temperature, humidity, and barometric pressure.
- Economically priced
- Thermometer, barometer, hygrometer
This economically priced 3 in 1 weather station provides convenient weather data in a simple yet classy manner. It measures temperature (Celsius), air pressure and air moisture levels.
Customers Who Bought This Also Bought