ContactUsLinkComponent
Eisco® Zinc Casted Slotted Masses, Set of 9 Weights, 100 g Total
Eisco® Zinc Casted Slotted Masses, Set of 9 Weights, 100 g Total
# de catalogue 470344-466
Fournisseur:  Wards
9 WGHTS1 HNGR EA10G
Eisco® Zinc Casted Slotted Masses, Set of 9 Weights, 100 g Total
# de catalogue 470344-466
Fournisseur:  Wards
Spécifications
  • Description:
    Zinc casted slotted masses, set of 9 weights, 100 g total
  • Cat. no.:
    470344-466
Spécifications
A propos de cet article
Set of 9 zinc-casted slotted masses with one hanger each of 10 g, total weight of 100 g. For use in physics experiments and demonstrations.
  • Powder-coated, die casted zinc
  • 10 g slotted weights with accuracy ±2%
  • Complete with hanger
  • Great alternative to expensive brass sets
Beautiful die casted zinc 9-piece slotted masses set on durable hanger. Each weight is 10 g, powder-coated, with a ±2% accuracy.
Customers Who Bought This Also Bought