Eisco® Zinc-Casted Slotted Mass Hanger, 10 to 100 g
Fournisseur: Wards
Certificats
A propos de cet article
Zinc-casted slotted mass hanger for use in physics experiments and demonstrations.
- 10, 50, or 100 g hangers designed for use with slotted mass sets
- Die-cast in zinc and powder-coated to ensure durability and long life
- A great financially-friendly alternative to expensive brass hangers
- Hanger has a ±2% accuracy
- Serves as a foundation for any set of 10 g slotted masses
These zinc-casted hangers are powder-coated for durability and serve as the foundation for any set of 10 g slotted masses. With a ±2% accuracy, they are an excellent alternate for expensive brass weights.
Customers Who Bought This Also Bought