Eisco® Connecting Leads with Alligator Clips
# de catalogue 470345-242
Fournisseur: Wards
Spécifications
- Description:Red connecting leads with alligator clips
- Cat. no.:470345-242
- Length:300 mm (12")
Spécifications
A propos de cet article
Red connecting leads, each 300 mm in length with alligator clips on both ends for 4 mm plug.
- 12" length (300 mm)
- Alligator clips at both ends
- Insulated
- Quick, temporary electrical connections
Customers Who Bought This Also Bought