Eisco® Economy Brass Hooked Weights, Set of 9
Catalog # 470344-054
Supplier: Wards
Specifications
- Description:Economy brass hooked weights, set of 9
- Cat. no.:470344-054
Specifications
About this item
Set of nine brass hooked weights ranging from 10 to 1000 g, perfect for classroom or lab experiments. Supplied in a wooden block for easy storage.
- 10 to 1000 g brass weights in a wooden block
- 7.75×3.75×3" wooden block for storage
As this is the economy set, the smaller seven weights are precisely machined, hooked, and made of brass. The two largest weights are a painted brass. All units are inscribed with their specific weight in grams. Wooden block included for storage. The set includes: (1) 1000 g , (1) 500 g, (2) 200 g, (1) 100 g, (1) 50 g, (2) 20 g, (1) 10 g.
Customers Who Bought This Also Bought