Eisco® Hooked Stainless Steel Weights, Set of 9
# de catalogue 470344-170
Fournisseur: Wards
Spécifications
- Description:Hooked stainless steel weights, set of 9
- Cat. no.:470344-170
Spécifications
A propos de cet article
Stainless steel hooked weights set with a capacity of 1000 g. Ideal for classroom or lab experiments.
- Stainless steel weights
- Premium wooden block
- Weights have hooks above and below main body, but can sit flat on a surface due to retractable bottom hook
- Will last for decades of lab and classroom use
Eisco Labs set of nine steel hooked weights, 10 to 1000 g in wooden block. Precisely machined hooked weights. Set contains (1) 1000 g, (1) 500 g, (2) 200 g, (1) 100 g, (1) 50 g, (2) 20 g, (1) 10 g.
Customers Who Bought This Also Bought