VWR® Adhesive Sealing Films for PCR and Robotics
Fournisseur: VWR International
Certificats
A propos de cet article
Features an extra strong adhesive to minimize evaporation, yet removes easily without leaving a residue on the plate.
- Certified DNase- and RNase-free
- Not recommended for qPCR
Standard cut features chamfered corner to eliminate plate overhang for robotics applications and tight storage conditions. Standard cut is ideal use with raised rim and non-skirted PCR plates.
Large cut design provides great plate coverage and easier positioning of the film on the plate. Large cut is recommended for use with flat-top, semi-skirted and full-skirted PCR plates as well as deep-well plates.
Customers Who Bought This Also Bought