VWR® General Purpose Sealing Films
Fournisseur: VWR International
Certificats
A propos de cet article
These general purpose seals are ideal for use with polypropylene plates. They minimize evaporation and adhesive plate residue.
- Certified DNase- and RNase-free
- Provides a very secure seal to minimize evaporation
- Can be used as a temporary seal while awaiting further processing
Provides superior barrier properties to minimize evaporation and prevent well to well contamination. The less rigid PP film reduces aerosols upon removal and has a specially formulated adhesive to minimize plate residue.
Customers Who Bought This Also Bought