PT1000 Dual Sensor
# de catalogue CA76777-186
Fournisseur: Heidolph NA, LLC
Spécifications
- Description:PT1000 dual sensor for monitoring temperature of sample and bath or heating block
- Cat. no.:CA76777-186
Spécifications
A propos de cet article
For monitoring temperature of sample and bath or heating block.
- Compatible with Hei-Plate Mix n Heat Expert and Ultimate models
Customers Who Bought This Also Bought