GC Accelerator Oven Insert Kit | Avantor
GC Accelerator Oven Insert Kit
GC accelerator oven insert kit, For Agilent 5890, 6890, 7890, and 8890 instruments
Catalog # 76204-502
Supplier:   Restek
Fournisseur #:  23849
GC Accelerator Oven Insert Kit
Catalog # 76204-502
Supplier:  Restek
GC Accelerator Oven Insert Kit
GC accelerator oven insert kit, For Agilent 5890, 6890, 7890, and 8890 instruments
Price
$2,427.44
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Designed with GC-MS users in mind, the GC Accelerator kit provides a simple way to speed up sample analysis.
  • Get the same GC separation in less time - use a GC Accelerator kit and the EZGC method translator to accurately convert methods to a scaled-down column format
  • Scaled-down methods let you speed up analysis time and increase sample throughput without capital investment
  • GC Accelerator kit installs easily without damaging the GC column or interfering with the MS interface
By reducing oven volume, these inserts allow faster ramp rates to be attained, which reduces oven cycle time and allows for increased sample throughput and more capacity to process rush samples. When faster ramp rates are used, existing methods can be accurately scaled down to smaller, high-efficiency, narrow-bore columns using Restek’s EZGC method translator.
With a scaled-down column, a properly translated method, and a GC Accelerator kit, you can obtain the same chromatographic separation often with greater sensitivity in a fraction of the time without making a capital investment.
Spécifications
  • Désignation:
    GC accelerator oven insert kit
  • For Use With:
    For Agilent 5890, 6890, 7890, and 8890 instruments
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.