VWR® Full Size Sliding Wire Basket for Select Refrigerator Units
Fournisseur: VWR International
Certificats
A propos de cet article
VWR® sliding storage baskets can streamline inventory management and enhance the safety and security of your products improving overall efficiency and productivity.
- Enhanced organization
- Improved air circulation
- Maximized storage space
Baskets are designed to fit 10.5 cu. ft. compact refrigerators and 5.2 cu. ft. freestanding undercounter refrigerators. Their sturdy construction allows you to stack and organize inventory for efficiency and easy accessibility.
Baskets are designed to promote superior air circulation, preventing hot spots and maintaining consistent temperatures throughout the chamber. This improves the shelf life of your products and reduces the risk of spoilage.
The bottom mounted fixed basket can add extra usable space to maximize your unit storage.
BodyTextAccessories: These VWR® Full size sliding wire baskets are compatible accessories for selected VWR® refrigerator units.
Ordering information: Full size baskets are factory installed only - See disclaimer.
Caution: Disclaimer: Full sized baskets are a factory-installed accessory. Bottom mounted baskets can be manually installed. The unit and the basket(s) must be ordered together. 10.5 cu. ft. units can replace up to 6 full sliding Baskets and 1 half-size, bottom-mounted fixed basket. 5.2 cu. ft. freestanding undercounter units can replace up to 3 full sliding Baskets and 1 half-size, bottom-mounted fixed basket. Lesser basket quantities can be ordered for each refrigerator and will be installed from the bottom of the unit upwards and equally spaced.
Customers Who Bought This Also Bought