Metal Lock Box | Avantor
Metal Lock Box
Generic printed metal lock box with 'Over Door' strap, 35.5×35.5×17.7 cm (14×14×7″)
Catalog # 76330-068
Supplier:   Therapak, LLC
Fournisseur #:  365172
Metal Lock Box
Catalog # 76330-068
Supplier:  Therapak, LLC
Metal Lock Box
Generic printed metal lock box with 'Over Door' strap, 35.5×35.5×17.7 cm (14×14×7″)
Price
$303.27
Log in pour la disponibilité
Each
Product Details & Documents
Each box is made with cold rolled steel for durability and powder-coated white for protection. It is insulated on the inside with foam to assist with preserving specimen integrity. A standard-keyed lock with 2 keys is provided with each unit.
  • Durable and made in the USA
  • Permanently attached straps
  • Customization available
Standard color: White
Standard printing: Reflex blue text, red biohazard symbol on front side only
Customization: Any deviation from the standard configuration, which may include changes in color, size, design features, closure and print.
Minimum quantity for custom print is 150 units.
Spécifications
  • Désignation:
    Generic printed metal lock box with 'Over Door' strap
  • Dimensions:
    35.5×35.5×17.7 cm (14×14×7″)