X-Plore® 8000 PAPR Sets
Fournisseur: Draeger
Certificats
A propos de cet article
An assortment of hood/helmet PAPR kits.
- Low maintenance; no calibration required
- Robust design with 10000 hour of expected life
- Superior comfort; guided air inlet optimizes air flow
- Optional shoulder harness provides superior comfort, ideal for long shifts
- Simple operation minimizes training
- Proprietary, intelligent alarm system with optical, acoustical, and vibration alarms
- RFID hose identification automatically adjusts for headpiece type
Heavy-duty design with industry leading IP 65 rating to protect against dust, airborne mist, and water vapor. High-capacity battery with an operating time of 16 to 20 hours on a single charge.
Workplace environments that compromise on the quality of air demand reliable solutions. The Dräger X-plore 8000 PAPR offers a new level of intuitive handling combined with intelligent electronics. The result is a high degree of personal protection needed to focus on the task at hand.
Certifications: IP 65 (dust-tight and resistant to water jets from all sides). NIOSH (42 CFR, part 84).
Ordering information: Filter units must be ordered separately.
Spécifications
- Catalog No:
- 76644-598
- 76644-600
- 76644-608
- Environmentally Preferable:
- Sustainable Packaging, Energy Efficient, Waste Reducing
- Sustainable Packaging, Energy Efficient, Waste Reducing
- Sustainable Packaging, Energy Efficient, Waste Reducing