CE Duo® CleanTech® Non Sterile Mops with Bucket Liner
# de catalogue 76635-846
Fournisseur: Vileda Professional - FHP
Spécifications
- Color:Blue
- Description:CE Duo® Cleantech® Nonsterile cleanroom foam mop
- Cat. no.:76635-846
- Mop Head Width:19.7"
Spécifications
A propos de cet article
This mop is now available in a conveinent six pack with a bucket liner. Cutting down not only on the time it takes to prepare the mops but also significantly reducing the amount of packaging waste.
- Microfiber, overstitched cleanroom foam for cleaning/disinfecting floors, walls, and ceilings
- Adhesive-free lamination techniqe prevents additional contamination
- Excellent microfiber cleaning results have been independently proven
- Reduce packaging waste
- Reduce amount of time needed to pre-prepare the mops
CE Duo® Cleantech® nonsterile mop is a single-use, double-sided microfiber cleanroom foam mop for cleaning and disinfecting floors, walls, and ceilings. The CE Duo® CleanTech® mop is packaged 6 mops per pack, single bagged with bucket liner.
Packaging: Mops are packaged 6 per pack, 8 packs per case.
Customers Who Bought This Also Bought